Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001107660.1 | NP_001101130.1 | 4 | 418 | |
XM_006232111.1 | XP_006232173.1 | 3 | 372 | ||
XM_008760842.1 | XP_008759064.1 | 4 | 375 | ||
GenBank mRNA | BC158888.1 | - | 4 | 381 | |
FQ225390.1 | - | 2 | 289 | ||
FQ225569.1 | - | 4 | 422 | ||
FQ225719.1 | - | 4 | 412 | ||
FQ231425.1 | - | 4 | 412 | ||
FQ233411.1 | - | 4 | 414 | ||
FQ235246.1 | - | 4 | 422 |