Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001267534.1 | NP_001254463.1 | 2 | 178 | |
XM_006234577.2 | XP_006234639.1 | 2 | 183 | ||
XM_006234578.2 | XP_006234640.1 | 2 | 312 | ||
XM_006234579.2 | XP_006234641.1 | 3 | 255 | ||
XM_006234580.2 | XP_006234642.1 | 2 | 272 | ||
XM_008762028.1 | XP_008760250.1 | 2 | 311 | ||
GenBank mRNA | BC083866.1 | - | 2 | 178 |