Gene Transcripts

Gene Symbol
Vom1r71
Entrez Gene ID
494243
Gene Name
vomeronasal 1 receptor 71
Gene Aliases
V1rc5
Chromosome Location
Chr. 4: 87608301 - 87609203 on Build Rnor_6.0
UniGene ID
Rn.129669
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009523.1 NP_001009523.1 1 756
GenBank mRNA AY510326.1 - 1 756

View Full Product Details