Gene Transcripts

Gene Symbol
Vom1r78
Entrez Gene ID
494273
Gene Name
vomeronasal 1 receptor 78
Gene Aliases
V1rc36
Chromosome Location
Chr. 4: 87943152 - 87944069 on Build Rnor_6.0
UniGene ID
Rn.129662
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008924.1 NP_001008924.1 1 626
GenBank mRNA AY510292.1 - 1 626

View Full Product Details