Gene Transcripts

Gene Symbol
Vom1r81
Entrez Gene ID
494313
Gene Name
vomeronasal 1 receptor 81
Gene Aliases
V1rc2
Chromosome Location
Chr. 4: 88048267 - 88049199 on Build Rnor_6.0
UniGene ID
Rn.129659
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008970.1 NP_001008970.1 1 591
GenBank mRNA AY510288.1 - 1 591

View Full Product Details