Gene Transcripts

Gene Symbol
Scgb2b24
Entrez Gene ID
494528
Gene Name
secretoglobin, family 2B, member 24
Gene Aliases
Abpbg3, Abpz
Chromosome Location
Chr. 1: 89778878 - 89779790 on Build Rnor_6.0
UniGene ID
Rn.137008
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_577783.2 XP_577783.2 2 136
GenBank mRNA AY871782.1 - 1 67
GU269242.1 - 2 136

View Full Product Details