Gene Transcripts

Gene Symbol
Arid1b
Entrez Gene ID
282546
Gene Name
AT rich interactive domain 1B (Swi1 like)
Gene Aliases
6A3-5, Smcf1, smcf-1
Chromosome Location
Chr. 1: 45923113 - 46230551 on Build Rnor_6.0
UniGene ID
Rn.214230
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008758744.1 XP_008756966.1 418
GenBank mRNA AI070578.1 - 374
CB793402.1 - 385

View Full Product Details