Gene Transcripts

Gene Symbol
Mypop
Entrez Gene ID
499090
Gene Name
Myb-related transcription factor, partner of profilin
Gene Aliases
P42pop, RGD1565160
Chromosome Location
Chr. 1: 79897706 - 79909336 on Build Rnor_6.0
UniGene ID
Rn.63885
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109139.1 NP_001102609.1 3 952
XM_006228452.2 XP_006228514.1 3 1951

View Full Product Details