Gene Transcripts

Gene Symbol
Scube2
Entrez Gene ID
499241
Gene Name
signal peptide, CUB domain, EGF-like 2
Gene Aliases
RGD1563998, Scube2-ps1
Chromosome Location
Chr. 1: 174428079 - 174495356 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003748970.3 XP_003749018.1 15 1865
XM_006230005.2 XP_006230067.1 14 1742

View Full Product Details