Gene Transcripts

Gene Symbol
Arhgef33
Entrez Gene ID
500608
Gene Name
Rho guanine nucleotide exchange factor (GEF) 33
Gene Aliases
RGD1560224
Chromosome Location
Chr. 6: 3012670 - 3100358 on Build Rnor_6.0
UniGene ID
Rn.215814
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008764441.1 XP_008762663.1 8 975
XM_008764442.1 XP_008762664.1 8 975
XM_008764443.1 XP_008762665.1 8 975
XM_008764444.1 XP_008762666.1 8 921
XM_008764445.1 XP_008762667.1 8 975
XM_008764446.1 XP_008762668.1 8 975
XM_008764447.1 XP_008762669.1 8 975
XM_008764448.1 XP_008762670.1 8 975
XM_008764449.1 XP_008762671.1 8 975
XM_008764450.1 XP_008762672.1 8 975

View Full Product Details