Gene Transcripts

Gene Symbol
Pja1
Entrez Gene ID
683077
Gene Name
praja ring finger 1, E3 ubiquitin protein ligase
Gene Aliases
Chromosome Location
Chr. X: 69213645 - 69218834 on Build Rnor_6.0
UniGene ID
Rn.190368
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001101006.1 NP_001094476.1 3 1239
XM_006257105.2 XP_006257167.1 2 2353
XM_006257106.2 XP_006257168.1 2 2070
XM_006257107.2 XP_006257169.1 3 1813
GenBank mRNA BC081885.1 - 3 1239

View Full Product Details