Gene Transcripts

Gene Symbol
Bend3
Entrez Gene ID
683923
Gene Name
BEN domain containing 3
Gene Aliases
Chromosome Location
Chr. 20: 48248201 - 48270905 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006256584.2 XP_006256646.1 4 1296
XM_008773024.1 XP_008771246.1 3 912

View Full Product Details