Gene Transcripts

Gene Symbol
Spx
Entrez Gene ID
691138
Gene Name
spexin hormone
Gene Aliases
Npq
Chromosome Location
Chr. 4: 176629973 - 176635698 on Build Rnor_6.0
UniGene ID
Rn.118301
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AY383715.1 - 218

View Full Product Details