Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001004271.1 | NP_001004271.1 | 1 | 651 | |
XM_008770060.1 | XP_008768282.1 | 1 | 678 | ||
GenBank mRNA | BC078782.1 | - | 1 | 651 | |
FQ219334.1 | - | 1 | 145 | ||
U06273.1 | - | 1 | 632 | ||
U06274.1 | - | 1 | 633 |