Gene Transcripts

Gene Symbol
MAP3K10
Entrez Gene ID
4294
Gene Name
mitogen-activated protein kinase kinase kinase 10
Gene Aliases
MEKK10, MLK2, MST
Chromosome Location
Chr. 19: 40191657 - 40215575 on Build GRCh38
UniGene ID
Hs.466743
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002446.3 NP_002437.2 4 1392
XM_006723222.2 XP_006723285.1 2 251
XM_011526981.2 XP_011525283.1 4 1503
XM_011526982.2 XP_011525284.1 4 1502
GenBank mRNA X90846.1 - 4 1392
Z48615.1 - 4 1264

View Full Product Details