Gene Transcripts

Gene Symbol
Mir7-1
Entrez Gene ID
723902
Gene Name
microRNA 7-1
Gene Aliases
Mirn7-1, mir-7a-1, mmu-mir-7-1
Chromosome Location
Chr. 13: 58392779 - 58392886 on Build GRCm38
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_029825.1 1 78


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AA881786.1 - 1 129 Chr. 13 - 58392382 - 58392937 [-]

View Full Product Details