Gene Transcripts

Gene Symbol
Snhg1
Entrez Gene ID
83673
Gene Name
small nucleolar RNA host gene 1
Gene Aliases
Chromosome Location
Chr. 19: 8723487 - 8726326 on Build GRCm38
UniGene ID
Mm.292489
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002896.3 4 171
GenBank mRNA AK036616.1 - 1 1207
AK043166.1 - 4 172
AK051045.1 - 2 993


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AA646653.1 - 3 140 Chr. 19 - 8724152 - 8726312 [+]
GenBankmRNA CF911094.1 - 1 294 Chr. 19 - 8724403 - 8725522 [+]
GenBankmRNA AV515710.1 - 3 150 Chr. 19 - 8724140 - 8726323 [+]
GenBankmRNA AV497308.1 - 3 151 Chr. 19 - 8724155 - 8726331 [+]
GenBankmRNA BF120608.1 - 2 320 Chr. 19 - 8724158 - 8726436 [+]
GenBankmRNA BI456898.1 - 3 154 Chr. 19 - 8724140 - 8726443 [+]
GenBankmRNA AV515526.1 - 2 111 Chr. 19 - 8724394 - 8725805 [+]
GenBankmRNA W20856.1 - 157 Chr. - - [-]
GenBankmRNA AV460418.1 - 3 154 Chr. 19 - 8724138 - 8726323 [+]
GenBankmRNA AA433008.1 - 3 155 Chr. 19 - 8724140 - 8726432 [+]
GenBankmRNA EL960934.1 - 3 158 Chr. 19 - 8724140 - 8726443 [+]
GenBankmRNA AA162697.1 - 3 145 Chr. 19 - 8724148 - 8726310 [+]
GenBankmRNA CB589630.1 - 132 Chr. - - [-]
GenBankmRNA BB640482.1 - 4 168 Chr. 19 - 8723490 - 8726328 [+]
GenBankmRNA AA038088.1 - 3 155 Chr. 19 - 8724146 - 8726105 [+]
GenBankmRNA BE448643.1 - 3 144 Chr. 19 - 8724148 - 8726344 [+]
GenBankmRNA U40654.1 - 1 1363 Chr. - - [-]
GenBankmRNA BG091353.1 - 3 115 Chr. 19 - 8724177 - 8726261 [+]
GenBankmRNA DV053423.1 - 190 Chr. - - [-]
GenBankmRNA AA419715.1 - 3 155 Chr. 19 - 8724140 - 8725832 [+]
GenBankmRNA BY113650.1 - 4 168 Chr. 19 - 8723490 - 8725526 [+]
GenBankmRNA BY012584.1 - 4 168 Chr. 19 - 8723490 - 8726092 [+]
GenBankmRNA BF608049.1 - 195 Chr. - - [-]
GenBankmRNA AV501764.1 - 3 130 Chr. 19 - 8724162 - 8725341 [+]
GenBankmRNA AL118113.1 - 3 164 Chr. 19 - 8724138 - 8726273 [+]
GenBankmRNA AL034970.1 - 3 164 Chr. 19 - 8724138 - 8726409 [+]
GenBankmRNA BY098836.1 - 1 20 Chr. 19 - 8724673 - 8726359 [+]
GenBankmRNA AA124951.1 - 3 143 Chr. 19 - 8724152 - 8725797 [+]
GenBankmRNA AA795219.1 - 1 17 Chr. 19 - 8724676 - 8726370 [+]
GenBankmRNA AA178616.1 - 3 118 Chr. 19 - 8724174 - 8726395 [+]
GenBankmRNA BY412213.1 - 1 105 Chr. 19 - 8724588 - 8726442 [+]
GenBankmRNA AV461499.1 - 3 148 Chr. 19 - 8724157 - 8726363 [+]
GenBankmRNA AV443985.1 - 2 337 Chr. 19 - 8724143 - 8725802 [+]
GenBankmRNA AA037960.1 - 3 144 Chr. 19 - 8724152 - 8726343 [+]
GenBankmRNA CX567991.1 - 3 158 Chr. 19 - 8724140 - 8726443 [+]
GenBankmRNA W42022.1 - 150 Chr. - - [-]
GenBankmRNA AV570392.1 - 3 135 Chr. 19 - 8724157 - 8726390 [+]
GenBankmRNA AW990131.1 - 3 143 Chr. 19 - 8724149 - 8726394 [+]
GenBankmRNA CB590342.1 - 2 349 Chr. 19 - 8724131 - 8725879 [+]
GenBankmRNA BM730535.1 - 3 138 Chr. 19 - 8724154 - 8726421 [+]
GenBankmRNA CX566533.1 - 3 286 Chr. 19 - 8724005 - 8726443 [+]
GenBankmRNA AV492597.1 - 3 157 Chr. 19 - 8724140 - 8726345 [+]
GenBankmRNA AA200918.1 - 1 26 Chr. 19 - 8724669 - 8726420 [+]
GenBankmRNA EL963394.1 - 3 182 Chr. 19 - 8724155 - 8725242 [+]
GenBankmRNA BY391063.1 - 1 16 Chr. 19 - 8724677 - 8726441 [+]
GenBankmRNA BY111985.1 - 4 168 Chr. 19 - 8723490 - 8725825 [+]
GenBankmRNA BY013100.1 - 4 168 Chr. 19 - 8723490 - 8725832 [+]
GenBankmRNA AV487936.1 - 3 122 Chr. 19 - 8724170 - 8726374 [+]
GenBankmRNA X61845.1 - 96 Chr. - - [-]
GenBankmRNA W42055.1 - 153 Chr. - - [-]
GenBankmRNA BY328777.1 - 4 168 Chr. 19 - 8723490 - 8725810 [+]
GenBankmRNA AV475699.1 - 2 310 Chr. 19 - 8724170 - 8725256 [+]
GenBankmRNA AA547560.1 - 3 120 Chr. 19 - 8724172 - 8726099 [+]
GenBankmRNA AV481080.1 - 3 135 Chr. 19 - 8724157 - 8726376 [+]
GenBankmRNA AV495277.1 - 3 127 Chr. 19 - 8724168 - 8725256 [+]
GenBankmRNA AV569303.1 - 3 128 Chr. 19 - 8724164 - 8726438 [+]
GenBankmRNA AL361596.1 - 3 169 Chr. 19 - 8724140 - 8726105 [+]
GenBankmRNA AA543872.1 - 1 15 Chr. 19 - 8724678 - 8726442 [+]
GenBankmRNA AA028518.1 - 2 70 Chr. 19 - 8724435 - 8726342 [+]
GenBankmRNA BQ416548.1 - 3 146 Chr. 19 - 8724146 - 8726443 [+]
GenBankmRNA AA684508.1 - 2 97 Chr. 19 - 8724408 - 8726310 [+]
GenBankmRNA BX525735.1 - 3 145 Chr. 19 - 8724146 - 8726398 [+]
GenBankmRNA AV445236.1 - 3 161 Chr. 19 - 8724157 - 8726345 [+]
GenBankmRNA AV577661.1 - 2 336 Chr. 19 - 8724142 - 8725245 [+]
GenBankmRNA AA220561.1 - 3 133 Chr. 19 - 8724159 - 8726387 [+]
GenBankmRNA EL962085.1 - 3 205 Chr. 19 - 8724140 - 8726432 [+]
GenBankmRNA BG090562.1 - 3 115 Chr. 19 - 8724177 - 8726313 [+]
GenBankmRNA AW985636.1 - 3 154 Chr. 19 - 8724140 - 8726383 [+]
GenBankmRNA BU841485.1 - 177 Chr. - - [-]
GenBankmRNA AL118412.1 - 3 144 Chr. 19 - 8724148 - 8726309 [+]
GenBankmRNA BX528587.1 - 3 156 Chr. 19 - 8724140 - 8726387 [+]
GenBankmRNA BY147788.1 - 4 168 Chr. 19 - 8723490 - 8725808 [+]
GenBankmRNA AV584788.1 - 2 102 Chr. 19 - 8724418 - 8726393 [+]
GenBankmRNA CF896937.1 - 3 144 Chr. 19 - 8724148 - 8726442 [+]
GenBankmRNA AV505845.1 - 3 172 Chr. 19 - 8724147 - 8726323 [+]
GenBankmRNA AA061982.1 - 3 175 Chr. 19 - 8724140 - 8725046 [+]
GenBankmRNA AA462480.1 - 3 155 Chr. 19 - 8724140 - 8726311 [+]
GenBankmRNA BY113223.1 - 4 168 Chr. 19 - 8723490 - 8726081 [+]
GenBankmRNA BE371262.1 - 125 Chr. - - [-]
GenBankmRNA AA155410.1 - 3 143 Chr. 19 - 8724152 - 8726271 [+]
GenBankmRNA BF608050.1 - 3 156 Chr. 19 - 8724147 - 8726443 [+]
GenBankmRNA AV481116.1 - 3 134 Chr. 19 - 8724158 - 8726391 [+]
GenBankmRNA BQ257371.1 - 1 346 Chr. 19 - 8724356 - 8725303 [+]
GenBankmRNA BF607556.1 - 3 208 Chr. 19 - 8724140 - 8726443 [+]
GenBankmRNA BP427963.1 - 1 27 Chr. 19 - 8724669 - 8726443 [+]
GenBankmRNA BY098844.1 - 4 170 Chr. 19 - 8723488 - 8725823 [+]
GenBankmRNA CA542754.1 - 2 332 Chr. 19 - 8724148 - 8725802 [+]
GenBankmRNA AV501497.1 - 3 135 Chr. 19 - 8724157 - 8726389 [+]
GenBankmRNA BB254596.2 - 4 211 Chr. 19 - 8723487 - 8726326 [+]
GenBankmRNA BY114991.1 - 4 168 Chr. 19 - 8723490 - 8725519 [+]
GenBankmRNA AA110623.1 - 3 122 Chr. 19 - 8724170 - 8725927 [+]
GenBankmRNA AA760260.1 - 2 88 Chr. 19 - 8724140 - 8726396 [+]

View Full Product Details