Gene Transcripts
- Gene Symbol
- Bambi-ps1
- Entrez Gene ID
- 81913
- Gene Name
- BMP and activin membrane-bound inhibitor, pseudogene (Xenopus laevis)
- Gene Aliases
- Chromosome Location
- Chr. 2: 122466583 - 122467797 on Build GRCm38
- UniGene ID
- Mm.122640
- Species
- Mus musculus
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BF720956.1 |
- |
1 |
478 |
|
Chr. 2 - 122467275 - 122467797 [-] |
GenBankmRNA |
AI645332.1 |
- |
1 |
478 |
|
Chr. 2 - 122467294 - 122467797 [-] |
View Full Product Details