Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NR_002710.2 | 14 | 2425 | ||
NR_120453.1 | 13 | 1872 | |||
GenBank mRNA | AL832768.1 | - | 14 | 2142 | |
BC041851.1 | - | 16 | 2606 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AA411551.1 | - | 4 | 468 | Chr. 17 - 6898868 - 6900094 [+] |