Gene Transcripts
- Gene Symbol
- SNORA10
- Entrez Gene ID
- 574042
- Gene Name
- small nucleolar RNA, H/ACA box 10
- Gene Aliases
- ACA10, SNORA10A
- Chromosome Location
- Chr. 16: 1962334 - 1962466 on Build GRCh38
- UniGene ID
- Hs.679928
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002327.1 |
|
1 |
111 |
|
GenBank mRNA |
AJ609432.1 |
- |
1 |
111 |
|
View Full Product Details