Gene Transcripts
- Gene Symbol
- C3orf49
- Entrez Gene ID
- 132200
- Gene Name
- chromosome 3 open reading frame 49
- Gene Aliases
- Chromosome Location
- Chr. 3: 63819365 - 63848636 on Build GRCh38
- UniGene ID
- Hs.506386
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026866.1 |
|
1 |
133 |
|
GenBank mRNA |
BC015210.1 |
- |
1 |
102 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BE790096.1 |
- |
|
80 |
|
Chr. - - [-] |
GenBankmRNA |
CV023702.1 |
- |
1 |
26 |
|
Chr. 3 - 63819472 - 63831177 [+] |
GenBankmRNA |
CD636568.1 |
- |
|
419 |
|
Chr. - - [-] |
GenBankmRNA |
AA448918.1 |
- |
1 |
75 |
|
Chr. 3 - 63819423 - 63823485 [+] |
GenBankmRNA |
BG198837.1 |
- |
1 |
137 |
|
Chr. 3 - 63819362 - 63827726 [+] |
View Full Product Details