Gene Transcripts
- Gene Symbol
- C3orf49
- Entrez Gene ID
- 132200
- Gene Name
- chromosome 3 open reading frame 49
- Gene Aliases
- Chromosome Location
- Chr. 3: 63819365 - 63848636 on Build GRCh38
- UniGene ID
- Hs.506386
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026866.1 |
|
7 |
1037 |
|
GenBank mRNA |
BC015210.1 |
- |
7 |
1006 |
|
View Full Product Details