Gene Transcripts

Gene Symbol
ANKRD26P1
Entrez Gene ID
124149
Gene Name
ankyrin repeat domain 26 pseudogene 1
Gene Aliases
Chromosome Location
Chr. 16: 46469337 - 46569097 on Build GRCh38
UniGene ID
Hs.97414
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026556.1 14 3135
GenBank mRNA AK125968.1 - 2 230
BC070117.1 - 14 3135
BX648636.1 - 4 918
CR749347.1 - 7 1758

View Full Product Details