Gene Transcripts

Gene Symbol
PRSS30P
Entrez Gene ID
124221
Gene Name
protease, serine, 30 pseudogene
Gene Aliases
Disp, TMPRSS8, TMPRSS8P
Chromosome Location
Chr. 16: 2839573 - 2842751 on Build GRCh38
UniGene ID
Hs.434388
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026864.2 1 1378
GenBank mRNA AK122946.1 - 1 1265
BC042597.1 - 2 1293

View Full Product Details