Gene Transcripts
- Gene Symbol
- SMAD5-AS1
- Entrez Gene ID
- 9597
- Gene Name
- SMAD5 antisense RNA 1
- Gene Aliases
- DAMS, SMAD5AS, SMAD5OS
- Chromosome Location
- Chr. 5: 136129513 - 136134890 on Build GRCh38
- UniGene ID
- Hs.59666
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026763.1 |
|
1 |
552 |
|
GenBank mRNA |
AF071111.1 |
- |
1 |
552 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000297163.3 |
- |
1 |
552 |
|
Chr. 5 - 136129516 - 136134890 [-] |
View Full Product Details