Gene Transcripts

Gene Symbol
PCA3
Entrez Gene ID
50652
Gene Name
prostate cancer associated 3 (non-protein coding)
Gene Aliases
DD3, NCRNA00019, PCAT3
Chromosome Location
Chr. 9: 76764436 - 76787569 on Build GRCh38
UniGene ID
Hs.663766
Species
Homo sapiens
Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_015342.2
3 663
NR_132312.1
4 828
NR_132313.1
3 528
GenBank mRNA AF103907.1 - LIT1552
DD3
4 828
AY894120.1 -
3 661


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AF103908.3 - 1 2340 Chr. 9 - 76782136 - 76787569 [+]

View Full Product Details