Gene Transcripts
- Gene Symbol
- SND1-IT1
- Entrez Gene ID
- 27099
- Gene Name
- SND1 intronic transcript 1
- Gene Aliases
- C7orf54, NAG8, NSG-X
- Chromosome Location
- Chr. 7: 127997509 - 128000077 on Build GRCh38
- UniGene ID
- Hs.657377
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027330.1 |
|
1 |
2029 |
|
GenBank mRNA |
AK024176.1 |
- |
1 |
1941 |
|
View Full Product Details