Gene Transcripts

Gene Symbol
SND1-IT1
Entrez Gene ID
27099
Gene Name
SND1 intronic transcript 1
Gene Aliases
C7orf54, NAG8, NSG-X
Chromosome Location
Chr. 7: 127997509 - 128000077 on Build GRCh38
UniGene ID
Hs.657377
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027330.1 1 1587
GenBank mRNA AF095687.1 - 1 1292
AF191492.1 - 1 1587
AK024176.1 - 1 1499

View Full Product Details