Gene Transcripts
- Gene Symbol
- ZDHHC8P1
- Entrez Gene ID
- 150244
- Gene Name
- zinc finger DHHC-type containing 8 pseudogene 1
- Gene Aliases
- ZDHHC8P
- Chromosome Location
- Chr. 22: 23390605 - 23402612 on Build GRCh38
- UniGene ID
- Hs.386693
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003950.1 |
|
6 |
2014 |
|
GenBank mRNA |
AK092530.1 |
- |
5 |
1843 |
|
AK092581.1 |
- |
3 |
1603 |
|
View Full Product Details