Gene Transcripts
- Gene Symbol
- SCARNA9L
- Entrez Gene ID
- 100158262
- Gene Name
- small Cajal body-specific RNA 9-like
- Gene Aliases
- Chromosome Location
- Chr. X: 20136066 - 20136413 on Build GRCh38
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_023358.1 |
|
1 |
297 |
|
View Full Product Details