Gene Transcripts

Gene Symbol
RBM48
Entrez Gene ID
84060
Gene Name
RNA binding motif protein 48
Gene Aliases
C7orf64, HSPC304, MGC16142
Chromosome Location
Chr. 7: 92528709 - 92540481 on Build GRCh38
UniGene ID
Hs.147829
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032120.3 NP_115496.2 5 3600
GenBank mRNA BC007365.2 - 1 918

View Full Product Details