Gene Transcripts

Gene Symbol
TDRG1
Entrez Gene ID
732253
Gene Name
testis development related 1 (non-protein coding)
Gene Aliases
LINC00532
Chromosome Location
Chr. 6: 40378424 - 40379892 on Build GRCh38
UniGene ID
Hs.180197
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024015.1 1, 2 467
GenBank mRNA BC042123.1 - 1, 2 554
DQ168992.1 - 1, 2 467


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AA293846.1 - 1, 2 179 Chr. 6 - 40378711 - 40379597 [+]
GenBankmRNA BG183126.1 - 151 Chr. - - [-]
GenBankmRNA AA446148.1 - 2, 3 372 Chr. 6 - 40378369 - 40379419 [+]
GenBankmRNA AL045589.1 - 1, 2 228 Chr. 6 - 40378663 - 40379717 [+]
GenBankmRNA HY034178.1 - 2, 3 381 Chr. 6 - 40378337 - 40379335 [+]
GenBankmRNA BG701637.1 - 1, 2 559 Chr. 6 - 40378335 - 40379658 [+]
GenBankmRNA BU561761.1 - 1, 2 470 Chr. 6 - 40378423 - 40379457 [+]

View Full Product Details