Gene Transcripts

Gene Symbol
TDRG1
Entrez Gene ID
732253
Gene Name
testis development related 1 (non-protein coding)
Gene Aliases
LINC00532
Chromosome Location
Chr. 6: 40378424 - 40379892 on Build GRCh38
UniGene ID
Hs.180197
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024015.1 2 808
GenBank mRNA BC033995.2 - 2 895
BC042123.1 - 2 895
DQ168992.1 - 2 808

View Full Product Details