Gene Transcripts

Gene Symbol
MTMR9LP
Entrez Gene ID
339483
Gene Name
myotubularin related protein 9-like, pseudogene
Gene Aliases
MTMR9L
Chromosome Location
Chr. 1: 32231660 - 32241710 on Build GRCh38
UniGene ID
Hs.471067
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026850.1 4 1708
GenBank mRNA AK096109.1 - 3 1260
AL833686.1 - 4 1521
BC016171.1 - 2 664
BC041632.1 - 2 474


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AA251929.1 - 1 25 Chr. 1 - 32232052 - 32235028 [-]
GenBankmRNA CA775436.1 - 3 428 Chr. 1 - 32232158 - 32240056 [-]
GenBankmRNA AW967629.1 - 1 25 Chr. 1 - 32231747 - 32235028 [-]
GenBankmRNA BG761082.1 - 2 475 Chr. 1 - 32231912 - 32240232 [-]
GenBankmRNA CN430607.1 - 2 461 Chr. 1 - 32231943 - 32240219 [-]

View Full Product Details