Gene Transcripts

Gene Symbol
MTMR9LP
Entrez Gene ID
339483
Gene Name
myotubularin related protein 9-like, pseudogene
Gene Aliases
MTMR9L
Chromosome Location
Chr. 1: 32231660 - 32241710 on Build GRCh38
UniGene ID
Hs.471067
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026850.1 2 659
GenBank mRNA AK096109.1 - 1 211
AL833686.1 - 2 569


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BM906403.1 - 314 Chr. - - [-]
GenBankmRNA BM546796.1 - 2 600 Chr. 1 - 32240200 - 32241626 [-]
GenBankmRNA BI766582.1 - 2 621 Chr. 1 - 32240556 - 32241671 [-]
GenBankmRNA DA372791.1 - 2 360 Chr. 1 - 32240390 - 32241681 [-]
GenBankmRNA BX337429.2 - 2 574 Chr. 1 - 32240189 - 32241626 [-]
GenBankmRNA BM722964.1 - 1 51 Chr. 1 - 32240007 - 32240630 [-]
GenBankmRNA DR006902.1 - 2 277 Chr. 1 - 32240140 - 32241320 [-]
GenBankmRNA BX402486.2 - 2 573 Chr. 1 - 32240159 - 32241626 [-]
GenBankmRNA DA638824.1 - 1 213 Chr. 1 - 32240245 - 32240790 [-]

View Full Product Details