Gene Transcripts
- Gene Symbol
- SNORD97
- Entrez Gene ID
- 692223
- Gene Name
- small nucleolar RNA, C/D box 97
- Gene Aliases
- U97
- Chromosome Location
- Chr. 11: 10801467 - 10801608 on Build GRCh38
- UniGene ID
- Hs.723086
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_004403.1 |
|
1 |
33 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BQ431403.1 |
- |
1 |
13 |
|
Chr. 11 - 10800545 - 10801588 [-] |
View Full Product Details