Gene Transcripts

Gene Symbol
LINC00523
Entrez Gene ID
283601
Gene Name
long intergenic non-protein coding RNA 523
Gene Aliases
C14orf70
Chromosome Location
Chr. 14: 100657268 - 100672744 on Build GRCh38
UniGene ID
Hs.379802
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024096.1 1 92
GenBank mRNA AK094097.1 - 1 110
BX161445.1 - 1 92


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GENCODE ENST00000556697.1 - 1 110 Chr. 14 - 100657250 - 100672775 [+]
GENCODE ENST00000360899.2 - 1 92 Chr. 14 - 100657250 - 100672775 [+]

View Full Product Details