Gene Transcripts

Gene Symbol
APOO
Entrez Gene ID
79135
Gene Name
apolipoprotein O
Gene Aliases
FAM121B, MIC26, Mic23, My025
Chromosome Location
Chr. X: 23833348 - 23907940 on Build GRCh38
UniGene ID
Hs.495851
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026545.2 7, 8 805
XM_011545585.2 XP_011543887.1 8, 9 1169
XM_017029837.1 XP_016885326.1 8, 9 815
GenBank mRNA AF061264.1 - 7, 8 770
EL951748.1 - 4, 5 258

View Full Product Details