Gene Transcripts

Gene Symbol
ASZ1
Entrez Gene ID
136991
Gene Name
ankyrin repeat, SAM and basic leucine zipper domain containing 1
Gene Aliases
ALP1, ANKL1, C7orf7, CT1.19, GASZ, Orf3
Chromosome Location
Chr. 7: 117363222 - 117427523 on Build GRCh38
UniGene ID
Hs.352412
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC028695.1 - 11 1255
BC034963.1 - 11 1260 4824818
BC068488.1 - 10 1146

View Full Product Details