Gene Transcripts
- Gene Symbol
- SCARNA23
- Entrez Gene ID
- 677773
- Gene Name
- small Cajal body-specific RNA 23
- Gene Aliases
- ACA12
- Chromosome Location
- Chr. X: 24744441 - 24744570 on Build GRCh38
- UniGene ID
- Hs.689695
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003007.1 |
|
1 |
44 |
|
GenBank mRNA |
AJ609482.1 |
- |
1 |
44 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DW407923.1 |
- |
1 |
62 |
|
Chr. X - 24744439 - 24744879 [+] |
GenBankmRNA |
CN273799.1 |
- |
1 |
41 |
|
Chr. X - 24744452 - 24744880 [+] |
GenBankmRNA |
DW411968.1 |
- |
1 |
61 |
|
Chr. X - 24744440 - 24744749 [+] |
View Full Product Details