Gene Transcripts
- Gene Symbol
- SNORD15A
- Entrez Gene ID
- 6079
- Gene Name
- small nucleolar RNA, C/D box 15A
- Gene Aliases
- RNU15A, SNORNA, U15A
- Chromosome Location
- Chr. 11: 75400391 - 75400538 on Build GRCh38
- UniGene ID
- Hs.689081
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_000005.1 |
|
1 |
101 |
|
GenBank mRNA |
L15616.1 |
- |
1 |
101 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BI222262.1 |
- |
1 |
89 |
|
Chr. 11 - 75400404 - 75404156 [+] |
View Full Product Details