Gene Transcripts
- Gene Symbol
- SNORA53
- Entrez Gene ID
- 677832
- Gene Name
- small nucleolar RNA, H/ACA box 53
- Gene Aliases
- ACA53
- Chromosome Location
- Chr. 12: 98599635 - 98599884 on Build GRCh38
- UniGene ID
- Hs.693250
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003015.1 |
|
1 |
191 |
|
View Full Product Details