Gene Transcripts
- Gene Symbol
- ANKRD20A11P
- Entrez Gene ID
- 391267
- Gene Name
- ankyrin repeat domain 20 family member A11, pseudogene
- Gene Aliases
- C21orf81
- Chromosome Location
- Chr. 21: 13943769 - 13980444 on Build GRCh38
- UniGene ID
- Hs.364456
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027270.1 |
|
1 |
374 |
|
GenBank mRNA |
AF426257.1 |
- |
1 |
374 |
|
BC043199.1 |
- |
1 |
374 |
|
- Gene Symbol
- ANKRD20A5P
- Entrez Gene ID
- 440482
- Gene Name
- ankyrin repeat domain 20 family member A5, pseudogene
- Gene Aliases
- ANKRD20A5
- Chromosome Location
- Chr. 18: 14179097 - 14227050 on Build GRCh38
- UniGene ID
- Hs.632352
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_040113.1 |
|
1 |
115 |
|
GenBank mRNA |
BC022023.1 |
- |
1 |
115 |
|
DB446908.1 |
- |
1 |
90 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BG720199.1 |
- |
1 |
115 |
|
Chr. 18 - 14179100 - 14183738 [+] |
GenBankmRNA |
DB489467.2 |
- |
1 |
369 |
|
Chr. 21 - 13979778 - 13980439 [-] |
GenBankmRNA |
DB493896.2 |
- |
1 |
369 |
|
Chr. 21 - 13979712 - 13980439 [-] |
GenBankmRNA |
DB493485.2 |
- |
1 |
369 |
|
Chr. 21 - 13979724 - 13980439 [-] |
View Full Product Details