Gene Transcripts
- Gene Symbol
- PRCD
- Entrez Gene ID
- 768206
- Gene Name
- progressive rod-cone degeneration
- Gene Aliases
- RP36
- Chromosome Location
- Chr. 17: 76527586 - 76555339 on Build GRCh38
- UniGene ID
- Hs.634380
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_033357.1 |
|
1 |
172 |
|
GenBank mRNA |
AK125617.1 |
- |
1 |
172 |
|
View Full Product Details