Gene Transcripts

Gene Symbol
SNORD17
Entrez Gene ID
692086
Gene Name
small nucleolar RNA, C/D box 17
Gene Aliases
HBI-43
Chromosome Location
Chr. 20: 17962710 - 17962946 on Build GRCh38
UniGene ID
Hs.680747
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003045.1 1 204


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DA367252.1 - 197 Chr. - - [-]
GenBankmRNA BQ891750.1 - 1 179 Chr. 20 - 17962038 - 17962921 [-]
GenBankmRNA CB112228.1 - 1 203 Chr. 20 - 17962382 - 17962945 [-]
GenBankmRNA BF031032.1 - 184 Chr. - - [-]
GenBankmRNA DW409226.1 - 1 219 Chr. 20 - 17962623 - 17962956 [-]
GenBankmRNA BU945647.1 - 1 206 Chr. 20 - 17962372 - 17962946 [-]
GenBankmRNA CB157934.1 - 1 196 Chr. 20 - 17962659 - 17962938 [-]
GenBankmRNA DW408835.1 - 1 220 Chr. 20 - 17962622 - 17962946 [-]
GenBankmRNA BE865817.1 - 185 Chr. - - [-]
GenBankmRNA BU956161.1 - 187 Chr. - - [-]
GenBankmRNA BQ944667.1 - 1 179 Chr. 20 - 17961975 - 17962921 [-]
GenBankmRNA BM044438.1 - 1 181 Chr. 20 - 17962372 - 17962922 [-]
GenBankmRNA DA998121.1 - 1 201 Chr. 20 - 17962378 - 17962943 [-]
GenBankmRNA BU542144.1 - 1 179 Chr. 20 - 17962146 - 17962921 [-]
GenBankmRNA BM467639.1 - 1 179 Chr. 20 - 17962199 - 17962915 [-]
GenBankmRNA BG759456.1 - 1 190 Chr. 20 - 17962223 - 17962930 [-]
GenBankmRNA DA789110.1 - 196 Chr. - - [-]
GenBankmRNA BQ891736.1 - 1 179 Chr. 20 - 17962005 - 17962921 [-]
GenBankmRNA BG773728.1 - 1 185 Chr. 20 - 17962226 - 17962927 [-]

View Full Product Details