Gene Transcripts
- Gene Symbol
- LINC01548
- Entrez Gene ID
- 728409
- Gene Name
- long intergenic non-protein coding RNA 1548
- Gene Aliases
- C21orf54
- Chromosome Location
- Chr. 21: 33165470 - 33170236 on Build GRCh38
- UniGene ID
- Hs.125234
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024102.1 |
|
4 |
484 |
|
GenBank mRNA |
BX116511.1 |
- |
4 |
437 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AA400173.1 |
- |
4 |
437 |
|
Chr. 21 - 33165535 - 33170189 [-] |
View Full Product Details