Gene Transcripts

Gene Symbol
B3GALT5-AS1
Entrez Gene ID
114041
Gene Name
B3GALT5 antisense RNA 1
Gene Aliases
C21orf88
Chromosome Location
Chr. 21: 39597147 - 39612822 on Build GRCh38
UniGene ID
Hs.375120
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026542.1 1 212
NR_026543.1 1 212
GenBank mRNA BC062296.1 - 1 214
BC080530.1 - 1 211
BG751912.1 - 1 212


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BU174860.1 - 1 213 Chr. 21 - 39606118 - 39612823 [-]

View Full Product Details