Gene Transcripts
- Gene Symbol
- TTC3P1
- Entrez Gene ID
- 286495
- Gene Name
- tetratricopeptide repeat domain 3 pseudogene 1
- Gene Aliases
- RNF105L, TTC3L
- Chromosome Location
- Chr. X: 75740538 - 75743079 on Build GRCh38
- UniGene ID
- Hs.567832
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_030737.1 |
|
1 |
2519 |
|
GenBank mRNA |
AY672106.1 |
- |
1 |
365 |
|
View Full Product Details