Gene Transcripts

Gene Symbol
STUM
Entrez Gene ID
375057
Gene Name
stum, mechanosensory transduction mediator homolog
Gene Aliases
C1orf95
Chromosome Location
Chr. 1: 226548238 - 226609214 on Build GRCh38
UniGene ID
Hs.116827
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003665.3 NP_001003665.1 4 6087
XM_011544183.2 XP_011542485.1 3 6640
GenBank mRNA AF035308.1 - 1 1635


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BM692590.1 - 1 349 Chr. 1 - 226607248 - 226607734 [+]
GenBankmRNA DA175142.1 - 1 453 Chr. 1 - 226607144 - 226607693 [+]
GenBankmRNA DA172833.1 - 1 367 Chr. 1 - 226607230 - 226607789 [+]
GenBankmRNA DA360594.1 - 1 514 Chr. 1 - 226607084 - 226607627 [+]
GenBankmRNA DB177722.1 - 1 450 Chr. 1 - 226607148 - 226607689 [+]
GenBankmRNA DB179550.1 - 1 489 Chr. 1 - 226607109 - 226607658 [+]

View Full Product Details