Gene Transcripts
- Gene Symbol
- TCF12
- Entrez Gene ID
- 6938
- Gene Name
- transcription factor 12
- Gene Aliases
- CRS3, HEB, HTF4, HsT17266, TCF-12, bHLHb20
- Chromosome Location
- Chr. 15: 56918090 - 57291261 on Build GRCh38
- UniGene ID
- Hs.511504
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK056961.1 |
- |
1 |
845 |
|
View Full Product Details